Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formate Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB111748-100TEST
10186-932EA 1088.4 USD InStock
10186-932
Formate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Applications- The assay can detect formate in the range from 0.5 to 10 nmols/well.

Formic acid (HCOOH) is the simplest carboxylic acid and occurs naturally, notably in ant and bee venom. It is normally present at low levels in blood and urine (up to ~ 0.5 mM) but can be present in concentrations as high as 5 mM in acute methanol poisoning. Environmental exposure to formaldehyde can also elevate the blood and urine levels of formate. In the Formate Assay Kit, formate is oxidized to generate a product resulting in color formation (λ = 450 nm) proportional to formate concentration. The kit provides a convenient means for detecting formate in biological samples such as in serum or plasma, cells, culture and fermentation media. There is no need for pretreatment or purification of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty