Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citrate Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83396-100
10186-936EA 1061 USD InStock
10186-936
Citrate Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The Citrate Assay Kit can detect 0.1 to 10 nmoles (~2 µM-10 mM) of citrate in a variety of samples.

Citric acid (HOOC-CH₂-C(-OH)(-COOH)-CH₂-COOH) is a key intermediate in the TCA cycle which occurs in mitochondria. It is formed by the addition of oxaloacetate to the acetyl group of acetyl-CoA derived from the glycolytic pathway. Citrate can be transported out of mitochondria and converted back to acetyl CoA for fatty acid synthesis. Citrate is an allosteric modulator of both fatty acid synthesis (acetyl-CoA carboxylase) and glycolysis (phospho- fructokinase). Citrate is widely used industrially in foods, beverages and pharmaceuticals. Citrate metabolism and disposition can vary widely due to sex, age and a variety of other factors. BioVision's Citrate Assay Kit provides a simple, sensitive and rapid means of quantifying citrate in a variety of samples. In the assay, citrate is converted to pyruvate via oxaloacetate. The pyruvate is quantified by converting a nearly colorless probe to an intensely colored (570 nm) and fluorescent (Ex/Em, 535/587 nm) product. The Citrate Assay Kit can detect 0.1 to 10 nmoles (~2 µM-10 mM) of citrate in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty