Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isocitrate Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB83424-100
10186-938EA 511.5 USD InStock
10186-938
Isocitrate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- Isocitrate Assay Kit can detect 1 to 20 nmoles (~0.2 – 5 µg) of isocitrate.

Isocitric acid (HOOC-CHOH-CH (-COOH)-CH₂-COOH) is an intermediate of the Krebs TCA cycle, positioned between citrate and α-keto-glutarate. It is the branch point from which the glyoxylate shunt operates in plants and lower organisms. Isocitrate is found in substantial concentrations in many fruits and vegetables as well as in foods produced from these raw materials. In the TCA cycle, isocitrate is oxidized by isocitrate dehydrogenase (IDH) to α-ketoglutarate with the generation of NAD(P)H. Loss of NAD-IDH has been implicated as a potential causative factor in retinitis pigmentosa. BioVision's Isocitrate Assay Kit provides a simple, sensitive and rapid means of quantifying isocitrate in a variety of samples. In the assay, isocitrate is oxidized with the generation of NADPH which converts a nearly colorless probe to an intensely colored species with a λmax of 450nm. The Isocitrate Assay Kit can detect 1 to 20 nmoles (~0.2 – 5 µg) of isocitrate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty