Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxaloacetate Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83428-100
10186-944EA 675.66 USD InStock
10186-944
Oxaloacetate Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The Oxaloacetate Assay Kit can detect 0.1-10nmol (2-200 µM) of OAA.

Oxaloacetate (OAA, HOOC-CO-CH₂-COOH) is a TCA cycle intermediate. It precedes citrate which is formed by the transfer of an acetyl group to OAA. OAA is formed by the deamidation of aspartate or condensation of CO2 with pyruvate or PEP. Since mammals do not possess the enzymatic machinery to form TCA cycle intermediates from acetyl CoA, OAA is one of the anaplerotic entry points via pyruvate and pyruvate carboxykinase. BioVision's Oxaloacetate Assay Kit provides a simple, sensitive and rapid means of quantifying OAA in a variety of samples. In the assay, OAA is converted to pyruvate which is utilized to convert a nearly colorless probe to an intensely colored (λmax= 570nm) and fluorescent (Ex/Em = 535/587nm) product. The Oxaloacetate Assay Kit can detect 0.1-10nmol (2-200 µM) of OAA.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty