Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ascorbic Acid Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB65346-100
10186-946EA 530.37 USD InStock
10186-946
Ascorbic Acid Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The assay can detect 0.01-10 nmol of ascorbic acid per assay in various samples.

Ascorbic Acid (Vitamin C) plays an important role in many biological processes. It is a potent anti-oxidant, anti-inflammatory, anti-viral agent, and an immune stimulant and is present is a wide variety of foods and biological specimens. It is important to be able to monitor ascorbic acid content in these different samples. BioVision’s Ascorbic Acid Assay Kit provides a rapid, simple, and sensitive means of detecting ascorbic acid in various biological samples. In this assay, our proprietary catalyst oxidizes ascorbic acid to produce a product that interacts with the ascorbic acid probe, generating color and fluorescence. Ascorbic acid can be easily determined by either colorimetric (spectrophotometry at λ = 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The assay can detect 0.01-10 nmol of ascorbic acid per assay in various samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty