Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glucose Uptake Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB136956-100TEST
10186-948EA 1769.68 USD InStock
10186-948
Glucose Uptake Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Non-radioactive, Sensitive Assay

Detection method- Fluorescence (Ex/Em = 535/587 nm)
Species reactivity- Mammalian
Applications- This easy to use non-radioactive kit can detect glucose uptake as low as 50 pmol/well and can be used for a variety of cell types.

Glucose uptake is an important biological process for studying cell signaling and glucose metabolism. Among many different methods available for measuring glucose uptake, 2-deoxyglucose (2-DG) has been widely used because of its structural similarity to glucose. As with glucose, 2-DG can be taken up by glucose transporters, and metabolized to 2-DG-6-phosphate (2-DG6P). 2-DG6P, however, cannot be further metabolized, and thus accumulates in the cells. The accumulated 2-DG6P is, therefore, directly proportional to 2-DG (or glucose) uptake by cells. In BioVision’s glucose uptake fluorometric assay kit, the accumulated 2-DG6P is enzymatically oxidized and coupled to BioVision’s PicoProbe, which generates fluorescence in the presence of NADPH. This easy to use non-radioactive kit can detect glucose uptake as low as 50 pmol/well and can be used for a variety of cell types.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty