Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PicoProbe™ D-Lactate Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB174096-100TEST
10191-562EA 795.52 USD InStock
10191-562
PicoProbe™ D-Lactate Fluorometric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market

Detection method- Fluorescence (Ex/Em= 535/587 nm)
Applications:
-Measurement of D-Lactate in various tissues/cells/biological fluids
-Analysis of D-Lactate in pathological conditions
-Mechanistic study of the glyoxalase system

D-Lactate is the result of anaerobic glycolysis by microorganisms in the gastrointestinal system, and the product of detoxification of methylglyoxal by the glyoxalase system. The presence of abnormal levels of D-Lactate has been linked to a series of pathological conditions, such as diabetes, and appendicitis. BioVision’s PicoProbeTM D-Lactate Assay kit offers simplicity, sensitivity, and can be adapted to high-throughput research. The assay enzymatically oxidizes D-Lactate generating a fluorescent signal (Ex/Em = 535/587 nm). The signal is directly proportional to the amount of D-Lactate. The assay kit can detect samples containing as low as 0.1 µM D-Lactate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty