Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ceruloplasmin Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB273296-100TEST
10191-564EA 957.6 USD InStock
10191-564
Ceruloplasmin Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (OD 560 nm)
Applications:
- Measurement of ceruloplasmin activity in serum

Ceruloplasmin is a copper containing protein found primarily in the blood. It carries approximately 70% of the total copper present in the blood. Ceruloplasmin exhibits a weak oxidase activity, which is a more accurate method of assessing ceruloplasmin in serum than immunodiffusion or other non-enzymatic assays. Normal Ceruloplasmin levels are generally 1-4 µM (15-60 mg/dl), equivalent to approximately 50-150 mU/ml. Elevated amounts of serum ceruloplasmin are found in pregnancy, cancer, rheumatoid arthritis and in several mental conditions such as Alzheimer’s, schizophrenia and OCD. Abnormally low amounts are found in Wilson’s and Menkes’ Diseases and in several other rare conditions. BioVision’s Ceruloplasmin Activity Assay Kit utilizes an oxidase substrate, which gives an intensely colored product upon oxidation. The kit is fast, easy to use and suitable for high throughput applications and can measure ceruloplasmin activity in serum in the range between approximately 5 and 500 mU/ml.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty