Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ascorbic Acid Colorimetric Assay Kit (FRASC), BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB65656-100
10191-566EA 1083.02 USD InStock
10191-566
Ascorbic Acid Colorimetric Assay Kit (FRASC), BioVision
Assays Cellular Assays
Detection method- Absorbance (545-600 nm)
Species reactivity- Mammalian
Application- The assay can detect 0.2 to 20 nmol of ascorbic acid in various samples.

Ascorbic Acid (Vitamin C) plays an important role in many biological processes. It is a potent anti-oxidant, anti-inflammatory, anti-viral agent and an immune stimulant and is present in a wide variety of biological specimens. Due to the presence of a variety of other antioxidants in biological samples such as serum, most ascorbic acid assays show strong interference. BioVision’s FRASC Assay Kit provides a rapid, simple, and sensitive means of detecting ascorbic acid in biological samples such as serum and other body fluids, tissue and cell extracts, growth media and food products. In this assay, Feᶟᶧ is reduced to Fe²ᶧ by any antioxidants present. The ferrous iron is chelated with a colorimetric probe to produce a product with a strong absorbance band which can be monitored between 545-600 nm. The addition of ascorbate oxidase to parallel samples removes any ascorbate present leaving a background value which is subtracted from the total to give ascorbate content. The assay can detect 0.2 to 20 nmol of ascorbic acid in various samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty