Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alpha-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB185440-100TEST
10186-952EA 1409.63 USD InStock
10186-952
Alpha-Ketoglutarate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method-Absorbance (450 nm)
Applications- - Measurement of α-Ketoglutarate dehydrogenase activity in various tissues/cells
- Analysis of cell signaling pathways such as citrate acid cycle, lysine degradation or tryptophan metabolism in various cell types

α-Ketoglutarate Dehydrogenase (α-KGDH) (EC 1.2.4.2) is a key enzyme in the citric acid cycle. It forms an enzyme complex with dihydrolipoamide succinyl transferase (E2) and dihydrolipoamide dehydrogenase (E3). α-KGDH converts α-ketoglutarate into succinyl-CoA in the presence of NAD and CoA. It is highly regulated by intracellular ATP/ADP and NADH/NAD ratios and calcium. In humans, decreased KGDH activity can lead to neurodegenerative diseases such as Alzheimer’s disease. Recent studies show that α-KGDH is a target of oxidative stress; reactive oxygen species (ROS) inhibit KGDH activity which diminishes its critical function and can cause a bioenergetic deficit. BioVision’s α-KGDH assay kit provides a quick and easy way for monitoring α-KGDH activity in various samples. In the assay, α-KGDH converts α-ketoglutarate into an intermediate which reduces the probe to a colored product with strong absorbance at 450 nm. The assay is simple, sensitive and can detect α-ketoglutarate dehydrogenase activity lower than 0.1 mU in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty