Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvate Dehydrogenase (PDH) Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB287837-96TEST
10186-954EA 991.56 USD InStock
10186-954
Pyruvate Dehydrogenase (PDH) Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Applications- - Measurement of pyruvate dehydrogenase activity in various tissues/cells
- Analysis of cell signaling pathway

Pyruvate Dehydrogenase (PDH) (EC 1.2.4.1) has a vital role in carbohydrate metabolism. It forms a well-characterized enzyme complex with dihydrolipoyl transacetylase (E2) and dihydrolipoyl dehydrogenase (E3). PDH converts pyruvate into acetyl-CoA in the presence of NAD and CoA, and links glycolysis to the citric acid cycle. PDH activity is inhibited by high intracellular ratios of ATP/ADP, NADH/NAD or Acetyl-CoA/CoA. In humans, PDH deficiency reduces mitochondrial function and is linked to neurodegenerative diseases. PDH deficiency is X-linked; it results in 2 forms of abnormality: a metabolic form (lactic acidosis) and a neurological form (seizure and/or neuropathological spasm). Recent studies show that PDH is a target of oncogene-induced senescence; activation of PDH enhances pyruvate utilization and increases respiration and redox stress. BioVision’s PDH assay kit provides a quick and easy way for monitoring PDH activity in various samples. In the assay, PDH converts pyruvate into an intermediate, which reduces the developer to a colored product with strong absorbance at 450 nm. The assay is simple, sensitive and can detect pyruvate dehydrogenase activity lower than 0.1 mU in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty