Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PicoProbe™ Glucose-6-Phosphate Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB107923-100TEST
10186-958EA 995.9 USD InStock
10186-958
PicoProbe™ Glucose-6-Phosphate Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit can detect G6P in the range of 10 to 500 pmoles which is equivalent to the range of 1-500 µM in the original sample, in a high-throughput screening (HTS) format.

Glucose-6-phosphate (G6P) is a key intermediate for glucose transport into cells which then enters either metabolic pathways or storage. G6P can enter the glycolytic pathway, the pentose phosphate shunt or be stored as glycogen or starch. G6P is utilized by its dehydrogenase to generate reducing equivalents in the form of NAD(P)H. This is particularly important in red blood cells where a G6PDH deficiency leads to hemolytic anemia. BioVision's glucose-6-phosphate Assay Kit is a simple, sensitive and rapid means of quantifying G6P in a variety of samples. In the assay, glucose-6-phosphate is oxidized with the generation of a product that converts a nearly colorless probe to an intensely fluorescent product (Ex/Em 535/587 nm). The Glucose-6-phosphate Assay Kit can detect G6P in the range of 10 to 500 pmoles which is equivalent to the range of 1-500 µM in the original sample assuming a dilution of 5X during processing. The PicoProbe™ G6P Assay Kit is much more sensitive than the G6P Colorimetric Assay Kit for low G6P samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty