Would you like to log in again?
PicoProbe™ Glucose Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB169559-100TEST
10186-960EA 925.56 USD InStock
10186-960
PicoProbe™ Glucose Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS
Detection method- Fluorescence (Ex/Em = 535/587 nm)
Applications- This assay kit can detect less than 0.5 µM glucose in various biological samples. Analysis of metabolism and cell signaling. Mechanistic study of obesity and diabetes.
Glucose is the main energy source for virtually all living organisms. Glucose level is a key diagnostic parameter for many metabolic disorders. Measurement of glucose can be very important in both research and drug discovery processes. Biovision’s PicoProbe™ Glucose Assay kit is simple, rapid, ultra-sensitive and suitable for high-throughput. In this assay, D-glucose is enzymatically oxidized to form a product which reacts with a colorless probe to generate the fluorescence (Ex/Em = 535/587). The fluorescence generated is directly proportional to the amount of glucose. This assay kit can detect less than 0.5 µM glucose in various biological samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty