Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Glucosidase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB174093-100TEST
10186-964EA 912.56 USD InStock
10186-964
α-Glucosidase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive α-Glucosidase Activity Assay

Detection method- Absorbance (OD 410 nm)
Applications:
- Measurement of α-Glucosidase activity in biological samples
- Screening α-Glucosidase inhibitors

α-Glucosidase breaks down α-1,4 linked polysaccharides to glucose, which can be utilized as a source of energy. In the biotechnology industry, α-glucosidase is used to produce glucose from intermediate breakdown products of starch hydrolysis generated by enzymes such as amylase. Pompe disease, one of the 12 known glycogen storage diseases, is an autosomal recessive metabolic disorder attributed to α-glucosidase deficiency. In this disease, glycogen accumulates in the lysosomes, resulting in progressive muscle weakness, heart failure and other neurological symptoms. In BioVision’s α-Glucosidase Activity Colorimetric Assay Kit, α-Glucosidase hydrolyzes the Substrate Mix to release the p-nitrophenol that can be measured colorimetrically (OD = 410 nm). This is an easy, quick and high-throughput capable kit that can measure 0.1-10 mU of α-glucosidase activity in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty