Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lipase Activity Fluorometric Assay Kit III, BioVision

Supplier: BioVision
AB118969-100TEST
10186-984EA 881 USD InStock
10186-984
Lipase Activity Fluorometric Assay Kit III, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS

Detection method- Fluorescence (Ex/Em 529/600 nm)
Species reactivity- Mammalian
Applications- A sensitive and high speed homogeneous assay for detection of Lipase enzyme activity in a high-throughput screening (HTS) format.

Lipase is a subclass of the esterases that catalyze the hydrolysis of ester bonds of lipid substrates. Lipases perform essential roles in the digestion, transport and processing of dietary lipids (e.g. triglycerides, fats, oils) in most, if not all, living organisms. In humans, pancreatic lipase is the key enzyme responsible for breaking down fats in the digestive system by converting triglycerides to monoglycerides and free fatty acids. During the damage of the pancreas, lipase levels can rise 5 to 10-fold within 24 to 48 hours. In BioVision’s Lipase Assay Kit III, Lipase hydrolyzes a specific substrate to generate the methylresorufin, which can be detected fluorometrically at Em/Ex=529/600 nm. The kit provides a rapid, simple, more sensitive, and reliable test suitable for high throughput assay of Lipase activity. This kit can be used to detect Lipase as low as 0.1 mU/well.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty