Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDX60 Rabbit Polyclonal Antibody

Supplier: ProSci
6945_S
10751-692EA 185.24 USD InStock
10751-692
Anti-DDX60 Rabbit Polyclonal Antibody
Antibodies
DDX60 Antibody: DDX60 is a DEXD/H Box RNA helicase and the human homolog of the S. cerevisiae Ski2, a cofactor of RNA exosome. Expression of DDX60 increases after viral infection and binds to endogenous RIG-I, a cytoplasmic viral RNA sensor, as well as other RIG-I-like receptors such as MDA5 and LGP2. These proteins activate TBK1 and IKK-e, signaling IRF3 and NF-kB-responsive genes in the innate immune response pathway. DDX60 is required for the RIG-I- or MDA5-dependent response to viral infection, but is dispensable for TLR3-mediated signaling.

Type: Primary
Antigen: DDX60
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty