Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urokinase Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB174098-100TEST
10186-992EA 980.59 USD InStock
10186-992
Urokinase Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive Kit on the market

Detection method- Fluorescence (Ex/Em = 350/450 nm)
Application:
- Measuring Urokinase activity of purified enzymes
- Measuring Urokinase activity in serum & cell/nuclear extracts

Urokinase (urokinase-type plasminogen activator or uPA, EC 3.4.21.73) is a serine protease, which catalyzes the conversion of the plasminogen to active plasmin, which in turn participates in thrombolysis. Biovision’s Urokinase Fluorimetric Activity Assay Kit utilizes an AMC-based peptide substrate containing the recognition sequence for Urokinase. Urokinase present in the sample catalyzes the cleavage of the substrate and releases AMC, which can be easily quantified by measuring its fluorescence at Ex/Em = 350/450 nm. This sensitive assay detects very low levels of Urokinase activity in biological samples (as low as 0.02 IU/ml).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty