Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neuraminidase Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB185436-100TEST
10186-996EA 1049.54 USD InStock
10186-996
Neuraminidase Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- fluorescence (Ex/Em = 530/590 nm)
Application- Measurement of NA activity in various tissues/cells extracts or serum sample infected with influenza virus.

Neuraminidase (NA) is a very common enzyme that hydrolyzes terminal sialic acid residues on polysaccharide chains; most often a galactose residue. NA activity plays a key role in the invasion of target cells and the replication of influenza virus. NA activity also assists in the elution of progeny viruses from infected cells, and prevents self-aggregation of virus. Thus, NA is an important target for drug development. BioVision’s Neuraminidase Activity Assay Kit provides a simple and sensitive method for measuring NA activity using fluorescence (Ex/Em = 530/590 nm). The assay utilizes NA Probe to detect the neuraminidase activity. This high-throughput adaptable assay kit can detect NA activity as low as 2.0 mU/ml in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty