Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lipid Peroxidation (MDA) Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB118970-100TEST
10186-998EA 1138.64 USD InStock
10186-998
Lipid Peroxidation (MDA) Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method-Absorbance (532 nm) or Fluorescence (Ex/Em 532/553 nm)
Species reactivity- All
Applications- The kit detects MDA levels as low as 1 nmol/well colorimetrically and 0.1 nmol/well fluorometrically.

Quantification of lipid peroxidation is essential to assess oxidative stress in pathophysiological processes. Lipid peroxidation forms Malondialdehyde (MDA) and 4-hydroxynonenal (4-HNE), as natural bi-products. Measuring the end products of lipid peroxidation is one of the most widely accepted assays for oxidative damage. BioVision’s Lipid Peroxidation Assay Kit provides a convenient tool for sensitive detection of the MDA in a variety of samples. The MDA in the sample is reacted with Thiobarbituric Acid (TBA) to generate the MDA-TBA adduct. The MDA-TBA adduct can be easily quantified colorimetrically (λ = 532 nm) or fluorometrically (Ex/Em = 532/553 nm). This assay detects MDA levels as low as 1 nmol/well colorimetrically and 0.1 nmol/well fluorometrically.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty