Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myeloperoxidase (MPO) Colorimetric Activity Assay Kit, BioVision

Supplier: BioVision
AB105136-100TEST
10187-000EA 1163.82 USD InStock
10187-000
Myeloperoxidase (MPO) Colorimetric Activity Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (412 nm)
Species reactivity- Mammalian

Myeloperoxidase (MPO) is a peroxidase enzyme (EC 1.11.1.7) most abundantly expressed in neutrophil granulocytes. It is a lysosomal protein stored in the azurophilic granules of the neutrophil. MPO contains a heme pigment which causes its green color in secretions rich in neutrophils, such as pus and some forms of mucus. MPO catalyzes the production of hypochlorous acid (HClO) from hydrogen peroxide (H₂O₂) and chloride anion (Cl-, or the equivalent from a non-chlorine halide). MPO also oxidizes tyrosine to a tyrosyl radical using hydrogen peroxide as an oxidizing agent. In BioVision’s MPO Assay Kit, the HClO produced from H₂O₂ and Cl- is reacted with taurine to generate the taurine chloramine, which subsequently reacts with the TNB2- probe to eliminate color (λ = 412 nm). The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput activity assay of MPO. This kit can be used to detect MPO as low as 0.05 mU per well.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty