Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myeloperoxidase (MPO) Fluorometric Activity Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB111749-100TEST
10187-002EA 1070.9 USD InStock
10187-002
Myeloperoxidase (MPO) Fluorometric Activity Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS

Detection method- Fluorescence (Ex/Em = 485/525 nm)
Species reactivity- Mammalian

Myeloperoxidase (MPO) is a peroxidase enzyme (EC 1.11.1.7) most abundantly present in neutrophil granulocytes. It is a green hemoprotein found in neutrophils and monocytes that catalyzes the reaction of hydrogen peroxide and halide ions to form cytotoxic acids and other intermediates that play a role in the oxygen-dependent killing of tumor cells and microorganisms. Its heme pigment causes the green color in secretions rich in neutrophils, such as pus and some forms of mucus. Furthermore, it can oxidize tyrosine to a tyrosyl radical using hydrogen peroxide as an oxidizing agent. In BioVision’s MPO Assay Kit, MPO catalyzes the production of NaClO from H₂O₂ and NaCl. Subsequently, NaClO will react stoichiometrically with Aminophenyl fluorescein (APF) to generate fluorescein, which has a strong fluorescence and can be detected at Ex/Em = 485/525 nm. The kit provides a rapid, simple, sensitive, and reliable test suitable as a high throughput assay of MPO activity. This kit can be used to detect MPO activity as low as 0.5 μU per well.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty