Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aspartate Aminotransferase (AST or SGOT) Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB105135-100TEST
10187-006EA 929.86 USD InStock
10187-006
Aspartate Aminotransferase (AST or SGOT) Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- This assay detects AST/ SGOT with a detection limit of 10 mU per well.

Aspartate aminotransferase (AST), also known as Glutamate-oxaloacetate transaminase (GOT) is a transaminase (EC 2.6.1.1) similar to the more liver specific alanine transaminase (ALT). Although commonly included clinically as part of a diagnostic liver function test, AST has a broader clinical utility since it may also be elevated in diseases affecting other organs, such as the heart or muscles in myocardial infarction, also in acute pancreatitis, acute hemolytic anemia, severe burns, acute renal disease, musculoskeletal diseases and trauma. It catalyzes the reaction:
Aspartate + ⍺-Ketoglutarate ⇌ Oxaloacetate + Glutamate
Diagnostically, it is almost always measured in units/liter (U/L). In BioVision’s AST Assay Kit, an amino group is transferred of from aspartate to ⍺-ketoglutarate. The products of this reversible transamination reaction are oxaloacetate and glutamate. The glutamate is detected in a reaction that concomitantly converts a nearly colorless probe to color (λmax = 450 nm). The kit provides a rapid, simple, sensitive and reliable test suitable as a high throughput activity assay of AST with a detection limit of 10 mU per well.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty