Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arginase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB180877-100TEST
10187-010EA 952.15 USD InStock
10187-010
Arginase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market

Detection method- Absorbance (570 nm)
Applications- Measurement of Arginase Activity in various tissues/cells
- Analysis of urea cycle disorders

Arginase (EC 3.5.3.1) is the final enzyme of the Urea Cycle. It converts L-arginine into urea and L-ornithine and plays an important role in removing ammonium ion from the body. Arginase has two isoforms: Arginase I and Arginase II. Arginase I is mainly present in the liver and plays an important part in the urea cycle whereas Arginase II is present in kidney and other tissues and regulates Arginine/Ornithine concentration. Arginase deficiency can lead to severe symptoms including neurological impairment, dementia and hyperammonemia. Analysis of Arginase activity is fundamental to the study of the urea metabolic pathway. BioVision’s Arginase Activity Assay kit is simple, sensitive and rapid. In this assay, Arginase reacts with arginine & undergoes a series of reactions to form an intermediate that reacts stoichiometrically with OxiRed™ Probe to generate the colored product (OD 570 nm). The kit can detect Arginase activity less than 0.2 U/L in 96-well assay format.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty