Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Enteropeptidase/Enterokinase Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB170964-100TEST
10187-016EA 934.61 USD InStock
10187-016
Enteropeptidase/Enterokinase Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method - Fluorescence (Ex/Em = 380/500 nm)
Application:
- Measurement of Enteropeptidase activity in biological samples or purified Enteropeptidase activity.
- Removing tag from recombinant proteins having recognition motif.

Enteropeptidase (Enterokinase, EC 3.4.21.9) is a serine protease involved in activation of trypsinogen to trypsin, which in turn results in the activation of various digestive enzymes. It recognizes a highly specific amino acid sequence ‘DDDDK’ and cleaves after the lysine residue. High specific activity of Enteropeptidase has been utilized in cleaving a variety of native or fusion protein tags containing the above recognition motif. In Biovision’s Enteropeptidase Activity Assay Kit, we have utilized a peptide substrate containing the Enteropeptidase recognition sequence along with a fluorescent label ‘AFC’. Enteropeptidase catalyzes the cleavage of this substrate and releases the AFC molecule, which can be easily quantified by measuring its fluorescence at Ex/Em = 380/500 nm. This assay kit is simple and rapid and can detect Enteropeptidase activity as low as 1 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty