Would you like to log in again?
Enteropeptidase/Enterokinase Cleavage Kit, BioVision
Supplier: BioVision
AB207001-100TEST
10187-018EA 436 USD InStock
10187-018
Enteropeptidase/Enterokinase Cleavage Kit, BioVision
Assays Cellular Assays
Sufficient for cleaving at least 5 mg of target protein
Enteropeptidase (Enterokinase, EC 3.4.21.9) is a serine protease involved in activation of trypsinogen to trypsin, which in turn results in the activation of various digestive enzymes. It recognizes a highly specific amino acid sequence ‘DDDDK’ and cleaves after the lysine residue (K). The high specific activity of Enteropeptidase has been utilized in cleaving a variety of native or fusion proteins containing the above recognition motif. Biovision’s Enteropeptidase Cleavage Kit contains highly active light chain fragment of human Enteropeptidase. Our pure enzyme is an excellent tool to obtain a wild type protein sequence from a fusion protein, containing Enteropeptidase recognition sequence (DDDDK). This Enteropeptidase cleavage kit is sufficient for cleaving at least 5 mg of target protein. The residual enteropeptidase left in the reaction mix will not interfere with most of the downstream applications. Following cleavage of the target protein, Enteropeptidase can be removed using BioVision Hi-Bind Ni-QR Agarose beads.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty