Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphoglucomutase Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB155896-100TEST
10187-028EA 1149.85 USD InStock
10187-028
Phosphoglucomutase Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- The kit can detect phosphoglucomutase activity even less than 1 mU/reaction.

Phosphoglucomutase (PGM) plays a key role in carbohydrate metabolism and widely exists in all organisms. PGM interconverts Glucose-1-Phosphate (G1P) and Glucose-6-Phosphate (G6P) depending on the body requirement. When glycogen breaks down, G1P is generated and phosphoglucomutase converts G1P to G6P, which can go either to glycolytic pathway to generate ATP, or to pentose phosphate pathway to generate ribose and NADPH. On the other hand, when cells have extra energy, PGM converts G6P to G1P, which generates glycogen. In humans, phosphoglucomutase have 2 isoforms (PGM I and PGM II). PGM deficiency leads to glucose storage disease. Detection of abnormal phosphoglucomutase activity is crucial for diagnosis, prediction and treatment of the disease. In BioVision’s phosphoglucomutase assay, PGM converts glucose-1-phosphate to glucose-6-phosphate; the glucose-6-phosphate is oxidized by glucose-6-phosphate dehydrogenase to form NADH, which reduces a colorless probe to a colored product with strong absorbance at 450 nm. The Phosphoglucomutase Assay Kit is simple, sensitive and rapid and can detect PGM activity even less than 1 mU/reaction.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty