Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphoglucose Isomerase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB155897-100TEST
10187-030EA 772.56 USD InStock
10187-030
Phosphoglucose Isomerase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Absorbance (450 nm)
Sample Type- Mammalian, bacteria, yeast, fish etc.
Application- The kit can detect phosphoglucose isomerase activity less than 0.1 mU/reaction.

Phosphoglucose isomerase (PGI, EC 5.3.1.9) is an important housekeeping enzyme. PGI catalyzes the interconversion of glucose-6-phosphate to fructose-6-phosphate. PGI performs multiple functions & intracellularly plays key role in both glycolysis and gluconeogenesis. Extracellularly, PGI [also called Autocrine Motility Factor (AMF)] functions as a cytokine, which stimulates cell motility and is associated with tumor development and metastasis. In humans, PGI deficiency causes hemolytic anemia, whereas increased PGI activity is observed in many cancers such as gastrointestinal, kidney and breast cancer. Early detection of abnormal phosphoglucose isomerase activity is crucial for diagnosis, prediction and therapeutic strategy. In BioVision’s Phosphoglucose Isomerase Activity Assay Kit, PGI converts fructose-6-phosphate to glucose-6-phosphate; the glucose-6-phosphate is oxidized by glucose-6-phosphate dehydrogenase to form a product, which reacts with a colorless probe to give strong absorbance at 450 nm. The PGI assay is simple, sensitive and rapid and can detect phosphoglucose isomerase activity less than 0.1 mU/reaction.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty