Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Creatine Kinase (CK) Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB155901-100TEST
10187-034EA 1122.47 USD InStock
10187-034
Creatine Kinase (CK) Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Absorbance (450 nm)
Application- The kit can detect creatine kinase activity less than 1 mU.

Creatine Kinase (CK) also known as creatine phosphokinase (CPK) and ATP: creatine N-phosphotransferase is a common cellular enzyme (EC 2.7.3.2). It catalyzes the reversible conversion of creatine and ATP into ADP and phosphocreatine. CK is widely expressed in various tissues and cell types, with highest activity in striated muscles, heart tissue and brain. CK consists of two subunits: M (muscle) and B (brain), and has three isoenzymes: CK-MM (skeleton muscle), CK-MB (cardiac muscle), and CK-BB (brain). Increased CK level is associated with many diseases such as myocardial infarction, muscular dystrophy, pulmonary infarction and brain tumors. Accurate measurement of CK is crucial for early diagnosis, prediction and therapeutic strategy. In BioVision’s Creatine Kinase Activity Colorimetric Assay kit, creatine kinase converts creatine into phosphocreatine and ADP. The generated phoshocreatine and ADP reacts with CK Enzyme Mix to form an intermediate, which reduces a colorless Probe to a colored product with strong absorbance at 450 nm. The CK Activity Assay is high-throughput adaptable, simple and sensitive. This assay kit can detect Creatine Kinase activity less than 1 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty