Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protease Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB111750-100TEST
10187-038EA 799.83 USD InStock
10187-038
Protease Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Fluorescence (Ex/Em 485/530 nm)
Applications- This kit can detect < 500 pg/well Trypsin present in the sample.

Proteases are naturally present in all organisms. These enzymes are involved in a multitude of physiological reactions from simple digestion of food proteins to highly regulated cascades. Proteases can either break specific peptide bonds (limited proteolysis), depending on the amino acid sequence of a protein, or break down a complete peptide to amino acids (unlimited proteolysis). The activity can be a destructive change (abolishing a protein's function), an activation of a function (preform to mature form) or it can be a signal in a signaling pathway. BioVision’s Protease Activity Assay Kit is designed for the quantitative determination of proteases present in the protein sample. The assay uses fluorescein isothiocyanate (FITC)-labeled casein as a general protease substrate. The fluorescein label on the FITC-Casein is highly quenched. Upon digestion by proteases present in the sample the FITC-Casein substrate is cleaved into smaller peptides which abolishes the quenching of the fluorescence label. The fluorescence of the FITC-labeled peptide fragments is measured at Ex/Em = 485/530 nm. The kit is supplied with our Mass Spectrometry Grade (MSG), chemically stabilized Trypsin for use as a general protease control. However, other protease standard controls can also be used. This kit is easy to use and can detect < 500 pg/well Trypsin present in the sample.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty