Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MMP-3 Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB118972-100TEST
10187-042EA 1049.52 USD InStock
10187-042
MMP-3 Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay, HTS

Detection method- Fluorescence (Ex/Em 325/393 nm)
Species reactivity- Mammalian
Applications- The assay sensitivity is < 50 µU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty