Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gamma Glutamyl Transferase (GGT) Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB241030-100TEST
10187-046EA 743.09 USD InStock
10187-046
Gamma Glutamyl Transferase (GGT) Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Fluorescence (Ex/Em 365/460 nm)
Species reactivity- Mammalian
Application- This assay detects GGT activity as low as 0.02 mIU in sample.

The Gamma-GlutamylTransferase (GGT; EC 2.3.2.2) is an enzyme that transfers gamma-glutamyl functional groups. It is found in many tissues, the most notable one being the liver, and has significance in medicine as a diagnostic marker. BioVision’s Gamma-GlutamylTransferase Assay Kit provides a convenient tool for sensitive detection of the GGT in a variety of samples. The GGT in sample will recognize L-γ-Glutamyl-AMC as a specific substrate leading to proportional fluorescence development. The activity of GGT can be easily quantified fluorometrically (Ex/Em = 365/460 nm). This assay detects GGT activity as low as 0.02 mIU in sample.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty