Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glucose Dehydrogenase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB102532-100TEST
10187-048EA 995.89 USD InStock
10187-048
Glucose Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- This assay detects GDH activity as low as 0.01 mU with our unit definition.

Glucose 1-dehydrogenase (NADᶧ) (EC 1.1.1.118 ) is an enzyme that catalyzes the chemical reaction: D-glucose + NADᶧ↔D-glucono-1,5-lactone + NADH + H+. This enzyme belongs to the family of oxidoreductases, specifically those acting on the CH-OH group of donor with NADᶧ or NADPᶧ as acceptor. BioVision’s Glucose Dehydrogenase (GDH) Assay Kit provides a convenient tool for sensitive detection of the GDH in a variety of samples. The GDH present in sample will recognize D-glucose as a specific substrate leading to a proportional color development. The activity of GDH can be easily quantified colorimetrically (λ = 450 nm). This assay detects GDH activity as low as 0.01 mU with our unit definition.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty