Would you like to log in again?
Recombinant SDF1 alpha (from E. coli)
Supplier: BioVision
AB51939-10
10191-924EA 275.43 USD InStock
10191-924
Recombinant SDF1 alpha (from E. coli)
Proteins and Peptides
>99% Pure active Recombinant murine SDF-1alpha (CXCL12).
Determined by its ability to chemoattract human peripheral blood monocytes using a concentration range of 50-100 ng/ml.
Stromal Cell-Derived Factor-1α (SDF-1α) is a recently discovered protein belonging to the α-chemokine (C-X-C) family of cytokines. Recombinant murine SDF-1α is a 7.9 kDa protein containing 68 amino acids residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty