Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MMP-1 Inhibitor Screening Kit (Fluorometric), BioVision

Supplier: BioVision
AB118973-100TEST
10187-062EA 917.19 USD InStock
10187-062
MMP-1 Inhibitor Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Sensitive Assay to Screen MMP-1 Inhibitors, HTS

Detection method- Fluorescence (Ex/Em 490/520 nm)
Applications- The kit can be used to screen inhibitors of MMP-1.

The Matrix metalloproteinase-1 (MMP-1, Interstitial collagenase, fibroblast collagenase) is a member of a multigene family of calcium-dependent, zinc-containing endoproteinases, the matrix metalloproteinases (MMPs). The MMPs are responsible for the degradation of the extracellular matrix (ECM) including collagens, elastins, gelatin, matrix glycoproteins and proteoglycan during normal development and disease processes. MMPs are regulated by hormones, growth factors and cytokines. MMP-1 belongs to the subclass, the collagenases, and along with MMP-8, and MMP-13 are the only members of the MMP family that are capable of degrading the types I, II and III interstitial collagens with high efficiency. These collagens are primarily found in bone, cartilage and skin. In BioVision’s MMP-1 Inhibitor Screening Kit, MMP-1 hydrolyzes a specific FRET substrate to release a quenched fluorescent group, which can be detected at Ex/Em = 490/520nm. In presence of potent MMP-1 inhibitors the clevage of substrate will be inhibited or stopped. The kit provides a rapid, simple, sensitive, and reliable test suitable as a high throughput screening assay of MMP-1 inhibitors. For comparison of the relative efficacy of test inhibitors, a control inhibitor, GM 6001 (IC₅₀ = 0.4 nM for MMP-1) is included.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty