Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Renin Inhibitor Screening Kit (Fluorometric), BioVision

Supplier: BioVision
AB204723-100TEST
10187-064EA 972.91 USD InStock
10187-064
Renin Inhibitor Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Sensitive Assay to Screen Renin Inhibitors, HTS

Detection Method – Fluorescence (Ex/Em = 328/552 nm)
Application:
- Screening potential inhibitors of renin
- Characterize/study renin inhibitors

Renin (EC 3.4.23.15), also known as an angiotensinogenase, is an enzyme that participates in the renin-angiotensin system (RAS) which mediates extracellular volume (i.e. blood plasma, lymph and interstitial fluid), and arterial vasoconstriction. An over-active renin-angiotensin system leads to vasoconstriction and retention of sodium and water, causing hypertension. Renin inhibitors are widely used for the treatment of hypertension. BioVision’s Renin inhibitor screening Kit uses a synthetic peptide substrate with a fluorophore (EDANS) at one end and a quencher (DABCYL) at the other end. Renin catalyzes the cleavage of FRET substrate resulting in a product that is detected fluorometrically at Ex/Em = 328/552 nm. In the presence of a Renin inhibitor, the rate of hydrolysis of the substrate is decreased. The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput screening of Renin inhibitors also adaptable to a 384 well format.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty