Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Luciferase Reporter Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB287865-200TEST
10187-068EA 264.13 USD InStock
10187-068
Luciferase Reporter Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive Assay

Detection method- Luminescence
Species reactivity- Mammalian
Application- This Luciferase Reporter Assay Kit provides a simple means for detecting luciferase activity in transfected eukaryotic cells.

Firefly luciferase has been used as a sensitive reporter for studying gene regulation and function. Luciferase provides a 1000-fold increase in sensitivity in comparison to the standard chloramphenicol acetyltransferase (CAT) assay. Firefly luciferase catalyzes the oxidative carboxylation of luciferin, a reaction with the highest efficiency of any known bioluminescence reaction. The light emission from the reaction can be recorded using a luminometer. BioVision’s Luciferase Reporter Assay Kit provides a simple means for detecting luciferase activity in transfected eukaryotic cells. For measurement of expressed luciferase in vitro, luciferase is first extracted from transfected cells through cell lysis. Then CoA, ATP, Mg²ᶧ and buffer are added to the lysate (Inclusion of CoA yields a nearly constant light emission rather than typical flash Kinetics). The luminescent reaction is then triggered by an injection of luciferin, and the emitted light is recorded by a luminometer. By providing CoA, along with ATP, Mg²ᶧ in an optimized buffer solution, BioVision’s Luciferase Reporter Assay Kit ensure maximal sensitivity, consistent light output, as well as convenience and consistency when working with multiple samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty