Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beta-Galactosidase Staining Kit, BioVision

Supplier: BioVision
AB102534-250TEST
10187-070EA 550.19 USD InStock
10187-070
Beta-Galactosidase Staining Kit, BioVision
Assays Cellular Assays
Detection method- Microscopy (Light microscope, phase contrast microscope, fluorescent microscope)
Species reactivity- Mammalian
Application- The β-Galactosidase staining kit utilizes X-gal as the substrate and LacZ gene as the reporter for testing the efficiency of expression vector mediated gene transfer and for studying the regulation of promoters of genes.

The LacZ gene from E. coli is one of the most commonly used reporter genes for testing the efficiency of expression vector mediated gene transfer and for studying the regulation of promoters of genes. The LacZ gene encodes the enzyme β-galactosidase, which is very stable, resistant to proteolytic degradation, can utilize a variety of substrates and can be easily assayed in situ. The β-Galactosidase staining kit utilizes X-gal as the substrate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty