Would you like to log in again?
Carrez Clarification Reagent Kit, BioVision
Supplier: BioVision
AB202373-100TEST
10187-072EA 267.72 USD InStock
10187-072
Carrez Clarification Reagent Kit, BioVision
Assays Cellular Assays
For preparation of food samples for further analysis
Application- Unique tool to prepare food samples for further analysis (bioassay, HPLC etc.)
Carrez clarification reagents are used to treat a wide variety of samples intended to be analyzed by enzymatic or other means. The reagents cause precipitation of proteins and fats, elimination of interference due to a number of redox compounds, which can affect assays and eliminates turbidity and emulsions particularly in food testing. The treatment of samples with Carrez Clarification Reagents is simpler, less expensive and takes less time than other methods such as spin filtering. Use of Carrez reagents is not suitable for samples in which enzymatic activities are to be quantified. Most samples collected for analyses of small molecule analytes such as carbohydrates, alcohols, aldehydes and organic acids can be prepared using this reagent system. These reagents must not be used to prepare samples for analytes such as ascorbate, ammonia, citrate or analytes, which may be converted to these in enzyme based assays, such as urea (à ammonia), aconitate (à citrate). The procedure is quick, simple and dilutes samples by a minimal amount.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty