Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biotin Quantitation Kit (Colorimetric), BioVision

Supplier: BioVision
AB185441-100TEST
10187-076EA 566.12 USD InStock
10187-076
Biotin Quantitation Kit (Colorimetric), BioVision
Assays Protein Assays
The most sensitive kit on the market

Detection method- Absorbance (500 nm)
Application- Measuring free biotin or the substitution degree of Biotinylation (biotin labels) of proteins and antibodies

Biotin (Vitamin H) is an important biomolecule that has been widely used in Biotinylation reactions to label antibodies or other proteins of interest. Due to its low molecular weight, biotin usually does not cause any significant changes in protein conformation and biological activity. Biotinylated proteins/antibodies are widely isolated and assayed with streptavidin pull-down assay, affinity chromatography, ELISA and Western blotting etc. Biovision offers a variety of ready-to-use Biotinylation reagents (Cat. # 2346, 2345, 2347, 2322, 2326, 2323). It is often desired to determine the degree of substitution of such biotinylated biomolecules. Biovision’s Biotin Assay Kit is based on the differential binding of Streptavidin to Biotin and a dye, 2-(4-hydroxyazobenzene) benzoic acid (HABA). When HABA is bound to Streptavidin, addition of free biotin or biotinylated biomolecules results in its displacement in the reaction. These changes are manifested in terms of decrease in the overall absorption (OD 500 nm) and can be quantitatively correlated to the amount of biotin present in solution. The kit also provides Biotin as a Standard and Biotinylated BSA as a positive control. This kit can be used to estimate as low as 60 pmol or 15 ng of biotin in solution.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty