Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BCA Protein Quantitation Kit, BioVision

Supplier: BioVision
AB102536-1000TEST
10187-078EA 651.94 USD InStock
10187-078
BCA Protein Quantitation Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (562 nm)
Application- Assay is suitable for measuring protein concentration in the range of 0.5-30 μg protein (0.01-0.6 mg/ml).

The BCA Protein Assay Kit provides a simple, rapid, detergent tolerant (up to 5%) procedure for determining the concentration of proteins in solution. The method utilizes a copper (Cu²ᶧ) salt which can be reduced to the cuprous state by protein(s). The generated Cu²ᶧ ion forms an intensely colored complex with the bicinchoninic acid reagent with a very strong absorbance band centered at 562 nm. The intensity of the blue complex is proportional to the amount of protein in the sample. The BCA Protein Assay is suitable for measuring protein concentration in the range of 0.5-30 μg protein (0.01-0.6 mg/ml).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty