Would you like to log in again?
Extra Sense™ BCA Protein Assay Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB207002-2500TEST
10187-084EA 544.11 USD InStock
10187-084
Extra Sense™ BCA Protein Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market
Detection method- Absorbance (562 nm)
Detergent compatible
BioVision’s Extra Sense™ BCA Protein Assay kit provides an ultra-sensitive method to detect and quantify total protein concentration even in the presence of detergents. The Kit has been optimized for determining total protein content in low protein concentration samples (0.5-40 μg/ml). The assay is based on the reaction of bicinchoninic acid (BCA) with the cuprous cation (Cu+1), which is generated by reduction of cupric cation (Cu+2) with the protein in alkaline conditions. The Cu+1-BCA chelate is a water-soluble complex and exhibits a strong absorbance at 562 nm that is linear over a concentration range of 0.5-40 μg/ml. In general, protein concentrations are estimated with reference to a commonly used protein standard. The Kit includes Bovine Serum Albumin (BSA) as a protein standard for estimation of total protein content of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty