Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GFP Quantitation Kit, BioVision

Supplier: BioVision
AB235672-100TEST
10187-088EA 758.36 USD InStock
10187-088
GFP Quantitation Kit, BioVision
Assays Cellular Assays
Detection method- Fluorescence (Ex/Em 488/507 nm)
Species reactivity- Mammalian
Application- It detects a wide range of GFP concentration (0.01-10 ug/ml)

Green Fluorescent Protein (GFP) is a spontaneously fluorescent protein originally isolated from the jellyfish Aequorea Victoria (Chalfie et al, 1994). GFP has also been recombinant modified and enhanced (called EGFP). GFP and EGFP have been widely used as a valuable reporter molecule for in vivo visualization of gene expression events in a variety of cell types and organisms. Since GFP requires no additional substrates or cofactors, GFP fluorescence can be easily detected under fluorescence microscope. However, most imaging studies of GFP are only qualitative. Since quantitative analysis of GFP expression level in cells or tissues are more informative and have wide applications, BioVision has developed the GFP Quantification Kit which quantifies GFP in 96 micro-plate format. Cells or tissues can be homogenized directly in the GFP Assay Buffer. The quantity of GFP is determined by comparing its fluorescence with that of GFP standard. The kit can detect a wide range of GFP concentration (0.01-10 µg/ml). A GFP quench solution is also provided for determining auto-fluorescence of cell or tissue extracts. Each kit provides sufficient reagents to perform up to 100 assays, Ex/Em = 488/507 nm.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty