Would you like to log in again?
BCA Protein Assay Kit (Test Tube) - Reducing Agent Compatible, BioVision
Supplier: BioVision
AB207004-250TEST
10187-092EA 543.94 USD InStock
10187-092
BCA Protein Assay Kit (Test Tube) - Reducing Agent Compatible, BioVision
Assays Cellular Assays
Only kit on the market that is compatible with higher conc
Detection method- Absorbance (562 nm)
Reducing agent compatible
BioVision’s Reducing Agent Compatible BCA Protein Assay Kit is the only protein estimation kit on the market that is compatible with strong reducing agents such as TCEP up to 20 mM, DTT up to 10 mM & β-mercaptoethanol up to 35 mM. This kit is adapted from BioVision’s BCA Assay Kit II (cat. # K813) and is based on the chelation of bicinchoninic acid (BCA) with the cuprous cation (Cu+1), which is generated by reduction of cupric cation (Cu+2) with the protein in alkaline conditions. Reducing agents in samples interfere with BCA assay due to their ability to reduce Cu+2. BioVision’s Reducing Agent Compatible BCA Protein Assay Kit is useful for determining protein concentrations of samples containing DTT, TCEP & β-mercaptoethanol. The assay is linear over the widest range of protein concentration between 25-2000 μg/ml. In general, protein concentrations are calculated with reference to a commonly used protein standard. The kit also includes Bovine Serum Albumin (BSA) as a protein standard for estimation of total protein content of samples.
of reducing agents TCEP (20 mM), DTT (10 mM), β-ME (35 mM)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty