Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PAH (p-Aminohippuric Acid) Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB204725-100TEST
10187-094EA 772.57 USD InStock
10187-094
PAH (p-Aminohippuric Acid) Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (550 nm)
Species reactivity- Mammalian

p-Aminohippuric acid (PAH) is a derivative of hippuric acid and useful as a diagnostic agent for the measurement of renal plasma flow. About 20-30% is completely filtered by the glomerulus and not readsorbed by the tubules. The remainder which bypasses the glomerulus and enters the tubules is completely secreted. At low doses, PAH is almost completely removed with one-pass through the kidneys. Hence, the venous concentration of PAH is close to zero and PAH has been used to determine the effective renal plasma flow (eRPF) from the plasma. The venous concentration is usually <10% that of the plasma concentration so eRPF slightly underestimates the actual RPF. This error is generally accepted because of the ease with which PAH infusion allows calculation of eRPF. In the Assay, PAH reacts with Dimethylaminocinnamaldehyde (DACA) which gives a strongly colored derivative at 550 nm. The amounts of PAH in urine and plasma can easily be quantified.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty