Would you like to log in again?
Human Recombinant MIP3B (from E. coli)
Supplier: BioVision
AB285571-20UG AB285571-1MG
10191-938EA 12747.58 USD InStock
10191-938 10191-940
Human Recombinant MIP3B (from E. coli)
Proteins and Peptides
>99% Pure active Recombinant human MIP-3beta .
Determined by its ability to chemoattract human T cells using a concentration range of 10-50 ng/ml.
Human macrophage inflammatory protein-3 β (MIP-3 β, Exodus-3, ELC) is a member of the β-chemokine (C-C) family of cytokines. It plays an important role in the inflammatory response of human T cells and tissue macrophages. Recombinant human MIP-3 β is a 8.8 kDa protein containing 77 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty