Would you like to log in again?
Preadipocyte Isolation Kit, BioVision Inc.
Supplier: BioVision
AB196988-5G
10191-100EA 796.74 USD InStock
10191-100
Preadipocyte Isolation Kit, BioVision Inc.
Assays Cellular Assays
Only kit on the market to isolate preadipocytes from adipose tissue
  • Simple, rapid & convenient method to isolate preadipocytes from adipose tissue.
Application:
Isolation of preadipocytes from adipose tissue
Adipose tissue is loose connective tissue that accumulates in animals and serves primarily for energy storage, insulation, and thermoregulation. It is predominantly composed of adipocytes, but also contains a stromal vascular fraction consisting of preadipocytes, fibroblasts, vascular endothelial cells, and immune cells. Recent research has discovered a dynamic role of adipose tissue in the production and secretion of various hormones and as an active endocrine organ. The excessive accumulation of adipose is known as obesity. Ways to prevent over-accumulation of adipose is a field of intense research. In addition to obesity, adipose tissue has been found to participate in various physiological processes, including reproduction, angiogenesis, inflammation, cancer, and vascular homeostasis. BioVision’s Preadipocyte Isolation Kit provides the reagents and tools to isolate the stromal vascular fraction from adipose tissue. The resulting stromal vascular fraction is cultured on a tissue-culture plate, where preadipocytes adhere. The preadipocytes retain the ability to proliferate and differentiate into adipocytes when treated with differentiation-inducing components (Cat. # K579, 3T3-L1 Differentiation Kit). The preadipocytes can be used to study the process of adipogenesis and the differentiated adipocytes can be used to study lipolysis, endocrine activity, cell-signaling, and metabolic dysfunction.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty