Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HAT (H4) Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB239713-100TEST
10191-106EA 873.34 USD InStock
10191-106
HAT (H4) Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Sensitive assay to measure HAT activity

Detection method- Fluorescence (Ex/Em = 535/587 nm)
Applications- Measurement of HAT activity of immunoprecipitated samples
- Measurement of HAT activity of purified enzyme preparations
- Measurement of HAT activity in Nuclear Extracts.

Histone Acetyltransferases (HATs) are enzymes that acetylate histone substrates resulting in important regulatory effects on chromatin structure and assembly, and gene transcription. Modifications of these proteins by HATs play an important role in the control of gene expression, and their dysregulation has been linked to cancer, neurodegeneration, and other diseases. BioVision’s HAT Activity Assay Kit utilizes Acetyl CoA and H4 histone peptide as substrates. In this assay, HAT enzyme catalyzes the transfer of acetyl groups from Acetyl-CoA to the histone peptide, thereby generating two products - acetylated peptide and CoA-SH. The CoA-SH reacts with the developer and PicoProbeTM to generate a product that is detected fluorometrically at Ex/Em = 535/587 nm. The assay can detect HAT activity equivalent to as low as 1.5 µU of p300 in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty