Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lipid (Oil Red O) staining Kit, BioVision

Supplier: BioVision
AB287838-24TEST
10191-108EA 662.02 USD InStock
10191-108
Lipid (Oil Red O) staining Kit, BioVision
Assays Cellular Assays
Detection method- Microscopy.
Applications- Staining of neutral lipids in cells.

Lipid droplets are found in all eukaryotic organisms. They function as reservoirs of neutral lipids, primarily triacylglycerol and cholesterol esters. The stored lipids can be used for energy, steroid synthesis, or membrane formation. Lipid droplet accumulation is a normal function in cells. Adipocytes consist predominantly of lipid droplets; however excessive lipid droplet accumulation within cells can be an indicator of metabolic deficiency or pathogenesis. For example, excessive accumulation of lipids in liver cells (steatosis) can lead to cellular dysfunction. At the onset of atherosclerosis, macrophages engulf oxidized-LDL, develop into foam cells, and contribute to artery narrowing. BioVision’s Lipid (Oil Red O) Staining Kit allows selective detection of neutral lipids within cultured cells. The kit also includes Hematoxylin to stain nuclei. This kit provides enough reagents to stain two (2) 96-well plates, two (2) 6-well plates, or four (4) 100 mm culture dishes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty