Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

pCAF Inhibitor Screening Kit (Fluorometric)

Supplier: BioVision
AB196991-100TEST
10191-112EA 1041.81 USD InStock
10191-112
pCAF Inhibitor Screening Kit (Fluorometric)
Assays Cellular Assays
Simple method to screen pCAF Inhibitors

Detection method- Fluorescence (Ex/Em = 392/482 nm)
Applications- Screening/studying/characterizing potential inhibitors of pCAF

pCAF (P300/CBP-associated factor, EC 2.3.1.48) also known as K (lysine) acetyltransferase 2B (KAT2B), possesses intrinsic histone acetyltransferase (HAT) activity. pCAF contributes to transcriptional activation by acetylating chromatin and transcription factors through its HAT activity. Deregulated HAT activity plays an important role in various cancers. pCAF inhibitors have potential applications in cancer therapy. pCAF uses both histone 3 (H3) and histone 4 (H4) as substrates. BioVision’s pCAF Inhibitor Screening Kit utilizes H3 peptide and Acetyl CoA as the substrates. pCAF acetylates the peptide and generates Coenzyme A with a free thiol group. The CoA is detected using a Thiol Detecting Probe that reacts with thiol groups and gives enhanced fluorescence that can be measured at Ex/Em = 392/482 nm. In the presence of pCAF specific inhibitors, the enzymatic activity is inhibited resulting in decreased or total loss of fluorescence. This assay kit is a simple, sensitive, and rapid tool to screen potential pCAF inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty