Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Monoamine Oxidase B (MAO-B) Inhibitor Screening Kit (Fluorometric), BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB284511-100TEST
10191-124EA 965.26 USD InStock
10191-124
Monoamine Oxidase B (MAO-B) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Only kit on the market to screen monoamine oxidase B (MAO-B) inhibitors

Detection method: Fluorescence (Ex/Em = 535/587 nm)
Application:
Screening/studying/characterizing MAO-B inhibitors

Monoamine oxidases (MAO, EC 1.4.3.4) are a family of enzymes that can oxidize a wide variety of endogenous primary amines. Two isoforms, MAO-A and MAO-B, have been identified based on their substrate, inhibitor specificity, and tissue localization. MAO-B can oxidize primary amines, but its list of specific substrates (i.e. benzylamine, phenylethylamine) is more limited compared to MAO-A. MAO-B is a mitochondrial-bound enzyme that is ubiquitously expressed throughout the brain and other tissues. It has been investigated in numerous studies including Parkinson’s disease, Alzheimer’s, and tobacco addiction. Specific MAO-B inhibitors such as selegiline, & rasagiline have been used to treat Parkinson’s patients, but their benefits are considered rather modest. BioVision’s MAO-B Inhibitor Screening Kit offers a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of MAO-B inhibitors. The assay is based on the fluorometric detection of H2O2, one of the byproducts generated during the oxidative deamination of MAO substrate.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty